Skip to Content
MilliporeSigma
All Photos(1)

Documents

79271

Supelco

Trimethylsilyl methallylsulfinate

for GC derivatization, ≥90.0% (GC)

Synonym(s):

2-Methyl-2-propenesulfinic acid trimethylsilyl ester, SILMAS-TMS, Trimethylsilyl 2-methyl-2-propenesulfinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16O2SSi
CAS Number:
Molecular Weight:
192.35
Beilstein:
9043766
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

for GC derivatization

Assay

≥90.0% (GC)

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

storage temp.

2-8°C

SMILES string

CC(=C)CS(=O)O[Si](C)(C)C

InChI

1S/C7H16O2SSi/c1-7(2)6-10(8)9-11(3,4)5/h1,6H2,2-5H3

InChI key

AVMZLLSHIFLNSJ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silyl methallylsulfinates: efficient and powerful agents for the chemoselective silylation of alcohols, polyols, phenols and carboxylic acids.
Huang, X., et al.
Chemical Communications (Cambridge, England), 14, 1297-1299 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service