Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78820

Supelco

1-Phenyl-1H-tetrazole-5-thiol

for spectrophotometric det. of Pd, for the determination of Bi, ≥97.0%

Synonym(s):

1-Phenyl-5-mercaptotetrazole, 1-Phenyltetrazoline-5-thione, 5-Mercapto-1-phenyltetrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4S
CAS Number:
Molecular Weight:
178.21
Beilstein:
139068
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

≥97.0% (HPLC)
≥97.0%

form

powder

quality

for spectrophotometric det. of Pd
for the determination of Bi

technique(s)

UV/Vis spectroscopy: suitable

mp

145 °C (dec.) (lit.)

SMILES string

Sc1nnnn1-c2ccccc2

InChI

1S/C7H6N4S/c12-7-8-9-10-11(7)6-4-2-1-3-5-6/h1-5H,(H,8,10,12)

InChI key

GGZHVNZHFYCSEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Sens. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fresenius Journal of Analytical Chemistry, 261, 151-151 (1972)
Mohammad M Khodaei et al.
The Journal of organic chemistry, 73(7), 2527-2532 (2008-03-05)
In the presence of 1-phenyl-5-mercaptotetrazole as a nucleophile, electrochemical oxidations of 1,2- and 1,4-dihydroxybenzenes have been investigated in aqueous solution using cyclic voltammetry and controlled-potential coulometry. The voltammetric results indicate that an electrooxidative/Michael-type sequential reaction occurs between the mercaptide anion
J Kejha et al.
Ceskoslovenska farmacie, 39(7), 294-298 (1990-09-01)
For the investigation of new anti-inflammatory drugs, 1-phenyl-5-mercaptotetrazole (I) was selected as the principal structure. The purpose itself lay in an alkylation of the mercapto group with different remainders. In some cases at the same time a substituent was introduced
[Information from the Soviet Toxicology Center].
Gigiena truda i professional'nye zabolevaniia, (10)(10), 55-56 (1980-10-01)
K Nesmĕrák et al.
Folia microbiologica, 45(2), 138-142 (2001-03-29)
The antifungal effect of substituted 1-phenyl-5-mercaptotetrazoles was tested with Candida tropicalis, C. pseudotropicalis, C. mogii, Trichosporon cutaneum, Cryptococcus albidus and S. cerevisiae. Candida strains exhibited the lowest sensitivity to the compounds; the most sensitive was S. cerevisiae. The MIC values

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service