Recommended Products
grade
for microscopy
purum p.a.
Quality Level
form
liquid
impurities
≤0.1% aromatic compounds (as benzene) (UV)
refractive index
n20/D 1.420
density
0.75 g/mL at 20 °C
Related Categories
Application
Non-aromatic solvent mixture of saturated aliphatic hydrocarbons (C9-C11)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
<-40.0 °F - closed cup
Flash Point(C)
< -40 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service