Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77766

Sigma-Aldrich

1-Phenoxy-2-propanol

technical, ≥80% (GC)

Synonym(s):

Propylene glycol phenyl ether, Propylene phenoxytol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CH(OH)CH3
CAS Number:
Molecular Weight:
152.19
Beilstein:
1941356
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥80% (GC)

refractive index

n20/D 1.523 (lit.)
n20/D 1.525

bp

243 °C (lit.)

solubility

water: soluble 15.1 g/L at 20 °C

density

1.064 g/mL at 20 °C (lit.)

SMILES string

CC(O)COc1ccccc1

InChI

1S/C9H12O2/c1-8(10)7-11-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3

InChI key

IBLKWZIFZMJLFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and identification of propylene glycol phenyl ethers.
A R SEXTON et al.
Journal of the American Chemical Society, 70(11), 3606-3606 (1948-11-01)
Polymerization of 1, 2-epoxypropane and 1, 2-epoxycyclohexane by diethylzinc-polyhydric phenol and/or phenol or 1-phenoxy-2-propanol as catalysts.
Kuran W and Listos T.
Macromolecular Chemistry and Physics, 195(2), 401-411 (1994)
J Greenman
Microbios, 39(156), 101-108 (1984-01-01)
Propionibacterium acnes was grown in continuous culture in the presence of propylene phenoxetol. At sub-lethal concentrations of this antimicrobial agent (0.025-0.1% w/v) steady-state growth conditions were achieved. In comparison with the control, cell biomass, maximum specific growth rates and levels
I K Hegna et al.
Annales de l'Institut Pasteur. Microbiology, 139(4), 473-483 (1988-07-01)
The bactericidal and fungicidal effects of five disinfectants and one combination of two disinfectants were tested using a modified Kelsey-Sykes method in which living microorganisms suspended in a sterilized yeast suspension ("dirty" conditions) and in sterile distilled water ("clean" conditions)
Russell C Wyeth et al.
Journal of neuroscience methods, 176(2), 121-128 (2008-09-24)
Anaesthesia is often used in neurophysiological, surgical, and neuroanatomical protocols. Several anaesthetics, including magnesium chloride, volatiles (halothane, etc.), and barbiturates, have been used in gastropod neurobiology. 1-Phenoxy-2-propanol (PP) is another anaesthetic option that has not yet been used extensively. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service