Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77078

Supelco

Pentylbenzene

analytical standard

Synonym(s):

n-Amylbenzene, 1-Phenylpentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)4CH3
CAS Number:
Molecular Weight:
148.24
Beilstein:
1904359
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.488 (lit.)
n20/D 1.488

bp

205 °C (lit.)

mp

−75 °C (lit.)

density

0.863 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

SMILES string

CCCCCc1ccccc1

InChI

1S/C11H16/c1-2-3-5-8-11-9-6-4-7-10-11/h4,6-7,9-10H,2-3,5,8H2,1H3

InChI key

PWATWSYOIIXYMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Pentylbenzene may be used as an analytical standard to investigate the extraction of pentylbenzene from high molar mass alkanes (C14 and C17) such as tetradecane and heptadecane by using N-methyl-2-pyrrolidone.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extraction of pentylbenzene from high molar mass alkanes (C14 and C17) by N-methyl-2-pyrrolidone
Fandary SM, et al.
The Journal of Chemical Thermodynamics, 38(4), 455-460 (2006)
Yi Min et al.
Journal of chromatography. A, 1356, 148-156 (2014-07-08)
In this work, 1.9 μm reversed-phase packing materials with superficially porous structure were prepared to achieve the rapid and high efficient separation of peptides and proteins. The silica particles were synthesized via three steps, nonporous silica particle preparation by a

Articles

A shortened review of fundamentals of Porous Graphitic Carbon (PGC) materials. What they are and how do they work as a stationary phase material in HPLC?.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service