Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75778

Supelco

Vincamine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3
CAS Number:
Molecular Weight:
354.44
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0%

optical activity

[α]/D +43.0±2°, c = 1 in pyridine

shelf life

limited shelf life, expiry date on the label

mp

232 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC[C@@]12CCCN3CCc4c(C13)n(c5ccccc45)[C@](O)(C2)C(=O)OC

InChI

1S/C21H26N2O3/c1-3-20-10-6-11-22-12-9-15-14-7-4-5-8-16(14)23(17(15)18(20)22)21(25,13-20)19(24)26-2/h4-5,7-8,18,25H,3,6,9-13H2,1-2H3/t18-,20+,21+/m1/s1

InChI key

RXPRRQLKFXBCSJ-GIVPXCGWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vincamine is a monoterpenoid indole alkaloid found in the leaves of Vinca minor L., belonging to the Apocynaceae family.[1]

Application

Vincamine may be used as an analytical reference standard for the determination of the analyte isolated from plant samples of Apocynaceae family by chromatography techniques.[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indentification of vincamine indole alkaloids producing endophytic fungi isolated from Nerium indicum, Apocynaceae
Na R, et al.
Microbiological research, 192, 114-121 (2016)
Determination of alkaloids in Catharanthus roseus and Vinca minor by high-performance liquid chromatography--tandem mass spectrometry
Liu J, et al.
Analytical Letters, 49(8) (2016)
Optimization of HPLC determination method for vincamine in Vinca minor L.
Hou E, et al.
Analytical Letters, 30(12), 2555-2560 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service