Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75230

Sigma-Aldrich

4-Methylumbelliferyl heptanoate

for fluorescence, ≥98.0% (TLC)

Synonym(s):

4-Methylumbelliferyl enanthate, Heptanoic acid 4-methylumbelliferyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O4
CAS Number:
Molecular Weight:
288.34
Beilstein:
1256801
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence

Assay

≥98.0% (TLC)

mp

41-42 °C (lit.)

solubility

DMF: soluble
alcohols: soluble

fluorescence

λex 365 nm; λem 445 nm in 0.1 M Tris pH 8.0 (lipase)

storage temp.

−20°C

SMILES string

CCCCCCC(=O)Oc1ccc2C(C)=CC(=O)Oc2c1

InChI

1S/C17H20O4/c1-3-4-5-6-7-16(18)20-13-8-9-14-12(2)10-17(19)21-15(14)11-13/h8-11H,3-7H2,1-2H3

InChI key

FFNBFZWIBOIPIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

suitable as fluorogenic substrate for lipases

Other Notes

Fluorogenic substrate for lipase

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorometric substrate for sulfatase and lipase.
G G Guilbault et al.
Analytical chemistry, 41(14), 2006-2009 (1969-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service