Skip to Content
MilliporeSigma
All Photos(1)

Documents

75028

Millipore

Shigella Broth Base

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein peptone, 20.0 g/L
dextrose, 1.0 g/L
polysorbate 80, 1.5 g/L
potassium dihydrogen phosphate, 2.0 g/L
potassium hydrogen phosphate, 2.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

food and beverages
sample preparation

microbiology

suitability

Shigella spp.

Application

Shigella Broth Base is used for the isolation and cultivation of Shigella species from food acc. ISO 21567:2004.

Components

Chemical Composition:
Formula adjusted and/or supplemented as required to meet performance criteria

Preparation Note

Suspend 31.5 grams in 1000 ml distilled water. Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes. Cool medium to 45-50°C and add novobiocin (500 μg/L) under aseptic conditions. Mix well and dispense in sterile test tubes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haram Park et al.
The EMBO journal, 39(11), e104150-e104150 (2020-04-30)
Alternative splicing regulates trans-synaptic adhesions and synapse development, but supporting in vivo evidence is limited. PTPδ, a receptor tyrosine phosphatase adhering to multiple synaptic adhesion molecules, is associated with various neuropsychiatric disorders; however, its in vivo functions remain unclear. Here, we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service