Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75018

Supelco

1-Hexanethiol

analytical standard

Synonym(s):

1-Mercaptohexane, Hexyl mercaptan, MCH, Mercaptan C6

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5SH
CAS Number:
Molecular Weight:
118.24
Beilstein:
1731295
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.448-1.452
n20/D 1.450

bp

150-154 °C (lit.)

mp

−81-−80 °C (lit.)

density

0.832 g/mL at 25 °C

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCS

InChI

1S/C6H14S/c1-2-3-4-5-6-7/h7H,2-6H2,1H3

InChI key

PMBXCGGQNSVESQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hexanethiol is an organic sulfur compound that belongs to the class of mercaptans. Mercaptans have a characteristic odor and are regarded as the major contributors to several foods and beverage aromas.

Application

1-Hexanethiol may be used as a surrogate standard for the determination of wine polyfunctional mercaptans by gas chromatography−negative ion mass spectrometry analysis of their pentafluorobenzyl derivatives. It may also be used as an analytical standard for the determination of the analyte in alcoholic beverages by simultaneous derivatization/extraction and liquid chromatography-high resolution mass spectrometry.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of volatile thiols in alcoholic beverages by simultaneous derivatization/extraction and liquid chromatography-high resolution mass spectrometry
Vichi S, et al.
Food Chemistry, 175(2), 401-408 (2015)
Laura Mateo-Vivaracho et al.
Journal of chromatography. A, 1146(2), 242-250 (2007-02-24)
A fast method for the determination of aroma-powerful polyfunctional thiols at nanogram per liter level has been developed and applied to wine. A small volume of wine (6 mL) was extracted with 1.5 mL of benzene containing four internal standards.
Hao Chen et al.
PloS one, 10(2), e0115482-e0115482 (2015-02-07)
Giant cell arteritis (GCA) and Takayasu's disease are inflammatory vasculitic syndromes (IVS) causing sudden blindness and widespread arterial obstruction and aneurysm formation. Glucocorticoids and aspirin are mainstays of treatment, predominantly targeting T cells. Serp-1, a Myxomavirus-derived serpin, blocks macrophage and
Chang Liu et al.
The Analyst, 139(21), 5460-5465 (2014-08-30)
Due to its low growth rate and its fastidious nature, Mycobacterium tuberculosis is difficult to identify. Its rapid and sensitive detection is, however, critical for the control of tuberculosis. Molecular biology, and more recently electrochemical technology, have been exploited for
Laura Mateo-Vivaracho et al.
Journal of chromatography. A, 1185(1), 9-18 (2008-02-09)
A fast method for the determination of aroma-powerful polyfunctional thiols at ng L(-1) level has been developed. Mercaptans are selectively retained in a 50mg solid-phase extraction cartridge and derivatization takes place in the same cartridge at room temperature (25 degrees

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service