Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74812

Sigma-Aldrich

Octamethylene diisocyanate

purum, ≥98.0% (GC)

Synonym(s):

1,8-Diisocyanatooctane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)8NCO
CAS Number:
Molecular Weight:
196.25
Beilstein:
1772257
MDL number:
UNSPSC Code:
12162002

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.455

SMILES string

O=C=NCCCCCCCCN=C=O

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service