Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74560

Sigma-Aldrich

L-Norleucine

≥99.0% (NT)

Synonym(s):

(S)-(+)-2-Aminohexanoic acid, (S)-2-Aminocaproic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
1721750
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥99.0% (NT)

optical activity

[α]20/D +23±1°, c = 5% in 5 M HCl

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

mp

>300 °C (lit.)

SMILES string

CCCC[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-2-3-4-5(7)6(8)9/h5H,2-4,7H2,1H3,(H,8,9)/t5-/m0/s1

InChI key

LRQKBLKVPFOOQJ-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

As internal standard in amino acid analysis

Other Notes

Non-essential amino acid.

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Gardner
Analytical biochemistry, 150(1), 174-177 (1985-10-01)
To assess quantitatively the merits of internal standardization, an amino acid mixture of known composition has been analyzed by conventional automated amino acid analysis before and after being subjected to total acid hydrolysis. Both the precision (reproducibility) and the accuracy
End-to-end aggregation of myosin.
H NODA et al.
Biochimica et biophysica acta, 58, 598-601 (1962-04-23)
Internal standards for amino acid analysis.
J F Riordan et al.
Methods in enzymology, 47, 31-40 (1977-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service