Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74095

Sigma-Aldrich

3-Nitro-L-tyrosine ethyl ester hydrochloride

purum, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N2O5 · HCl
CAS Number:
Molecular Weight:
290.70
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

purum

Assay

≥99.0% (AT)

optical activity

[α]20/D +3.3±0.3°, c = 2% in H2O

mp

~185 °C (dec.)

application(s)

peptide synthesis

SMILES string

Cl.CCOC(=O)[C@@H](N)Cc1ccc(O)c(c1)[N+]([O-])=O

InChI

1S/C11H14N2O5.ClH/c1-2-18-11(15)8(12)5-7-3-4-10(14)9(6-7)13(16)17;/h3-4,6,8,14H,2,5,12H2,1H3;1H/t8-;/m0./s1

InChI key

GITKQXFBNJTMRP-QRPNPIFTSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Modifies proteins via carboxylic group

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Desvages et al.
European journal of biochemistry, 105(2), 259-266 (1980-04-01)
3-Phosphoglycerate kinase is inactivated by 1-cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate and nitrotyrosine ethyl ester. The coupling of 1 mol nitrotyrosine/mol enzyme is sufficient to inactivate the protein completely. A weak protection against inactivation is observed with each substrate added separately. In contrast, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service