Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73930

Supelco

4-Hydroxy-3-nitroso-1-naphthalenesulfonic acid

for spectrophotometric det. of Co, Fe, ≥99.0%

Synonym(s):

2-Nitroso-1-naphthol-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7NO5S · 4H2O
CAS Number:
Molecular Weight:
325.29
Beilstein:
2136360
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.0% (TLC)
≥99.0%

quality

for spectrophotometric det. of Co, Fe

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤1%

mp

140-145 °C (dec.)

SMILES string

Oc1c(cc(c2ccccc12)S(O)(=O)=O)N=O

Other Notes

Determination of cobalt[1][2]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Motomizu et al.
Talanta, 29(2), 89-94 (1982-02-01)
The importance of selecting the most suitable counter-cation in the solvent extraction-spectrophotometric determination of an anionic metal chelate containing a sulphonic acid group is demonstrated and discussed. In the case of cobalt and 2-nitroso-1-naphthol-4-sulphonic acid (nitroso-NW acid), the most suitable
S. Motomizu et al.
Analytica Chimica Acta, 120, 267-267 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service