Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73705

Supelco

Delphinidin 3-O-β-D-glucoside chloride

analytical standard

Synonym(s):

Delphinidin 3-O-β-D-glucopyranoside chloride, Delphinidin 3-monoglucoside, Delphinin chloride, Myrtillin chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO12
CAS Number:
Molecular Weight:
500.84
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1=CC(O)=C(C=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C(C3=CC(O)=C(O)C(O)=C3)=[O+]4)C4=C1.[Cl-]

InChI

1S/C21H20O12.ClH/c22-6-15-17(28)18(29)19(30)21(33-15)32-14-5-9-10(24)3-8(23)4-13(9)31-20(14)7-1-11(25)16(27)12(26)2-7;/h1-5,15,17-19,21-22,28-30H,6H2,(H4-,23,24,25,26,27);1H/t15-,17-,18+,19-,21-;/m1./s1

InChI key

ZJWIIMLSNZOCBP-BTTVDUMLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jang Hoon Kim et al.
Bioorganic chemistry, 87, 803-809 (2019-04-13)
Eight anthocyanidin derivatives (1-8) were evaluated as potential inhibitors of the catalysis of α-glucosidase. Among them, compounds 4 and 8 had the highest levels of inhibitory activity at 100 μM (IC50 values of 14.4 ± 0.1 and 29.7 ± 1.2 μM) and acted in a dose-dependent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service