Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73527

Supelco

Elemolic acid, mixture of α and β

analytical standard

Synonym(s):

α,β-Elemadienolic acid, α,β-Elemic acid, 3α,β-Hydroxytirucallic acid, 3-Hydroxytirucalla-8,24-dien-21-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
Molecular Weight:
456.70
UNSPSC Code:
85151701

grade

analytical standard

Assay

≥92.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C\C(C)=C\CC[C@@H]([C@@H]1CC[C@]2(C)C3=C(CC[C@@]12C)[C@@]4(C)CCC(O)C(C)(C)[C@@H]4CC3)C(O)=O

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service