Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72800

Supelco

4-Nitrobenzaldehyde

for spectrophotometric det. of amino sugars, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CHO
CAS Number:
Molecular Weight:
151.12
Beilstein:
386796
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

quality

for spectrophotometric det. of amino sugars

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤0.05%

mp

103-106 °C (lit.)
104-106 °C

SMILES string

[O-][N+](=O)c1ccc(C=O)cc1

InChI

1S/C7H5NO3/c9-5-6-1-3-7(4-2-6)8(10)11/h1-5H

InChI key

BXRFQSNOROATLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the spectrophotometric det. of amino carbohydrates[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A. Nakamura et al.
Chemical & Pharmaceutical Bulletin, 17, 770-770 (1969)
Mikhail Yu Karganov et al.
Biological trace element research, 198(2), 567-574 (2020-03-08)
The objective of the present study was investigation of tissue trace element distribution in a streptozotocin model of DM1 in rats. DM1 was modeled in 2-month-old male Wistar rats (n = 30) using intraperitoneal injection of 45 mg/kg b.w. (STZ1) and 55 mg/kg b.w.
Jinmin Fan et al.
Chemical communications (Cambridge, England), (32)(32), 3792-3794 (2008-08-08)
Double-stranded DNA of natural origin can be used to facilitate nitro-aldol (or Henry) reaction in aqueous solution.
Jinmo Kim et al.
Analytical chemistry, 77(13), 4137-4141 (2005-07-01)
A chemical derivatization technique in time-of-flight secondary ion mass spectrometry (TOF-SIMS) has been developed to quantify the surface density of amine groups of plasma-polymerized ethylenediamine thin film deposited on a glass surface by inductively coupled plasma chemical vapor deposition. Chemical
Gang Wu et al.
The journal of physical chemistry. A, 112(5), 1024-1032 (2008-01-16)
We have used solid-state 17O NMR experiments to determine the 17O quadrupole coupling (QC) tensor and chemical shift (CS) tensor for the carbonyl oxygen in p-nitro-[1-(17)O]benzaldehyde. Analyses of solid-state 17O NMR spectra obtained at 11.75 and 21.15 T under both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service