Skip to Content
MilliporeSigma
All Photos(1)

Documents

72679

Supelco

1-Amino-1-deoxy-D-ribitol hydrochloride

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H13NO4 · HCl
CAS Number:
Molecular Weight:
187.62
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95% (TLC)

optical activity

[α]/D -11.0±3.0°, c = 1 in 0.1 M HCl

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

NC[C@H](O)[C@H](O)[C@H](O)CO.[H]Cl

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service