Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72255

Sigma-Aldrich

Bis(cyclopentadienyl)nickel(II)

purum, ≥97.0% (Ni)

Synonym(s):

Di(cyclopentadienyl)nickel(II), Nickelocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ni(C5H5)2
CAS Number:
Molecular Weight:
188.88
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

purum

Assay

≥97.0% (Ni)

form

solid

mp

171-173 °C (lit.)

storage temp.

2-8°C

SMILES string

[Ni].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Ni/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

FOOKRXHSBKEWSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service