Skip to Content
MilliporeSigma
All Photos(1)

Documents

71336

Sigma-Aldrich

Sodium tert-butyl carbonate

technical

Synonym(s):

tert-Butyl carbonate sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCOONa
CAS Number:
Molecular Weight:
140.11
Beilstein:
6715256
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical

SMILES string

[Na+].CC(C)(C)OC([O-])=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service