Skip to Content
MilliporeSigma
All Photos(1)

Documents

70998

Sigma-Aldrich

1-(2-Methoxyethyl)piperazine

purum, ≥98.0% (GC)

Synonym(s):

4-(2-Methoxyethyl)piperazine, N-(2-Methoxyethyl)piperazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O
CAS Number:
Molecular Weight:
144.21
Beilstein:
105098
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.4730 (lit.)
n20/D 1.473

bp

193-194 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

SMILES string

COCCN1CCNCC1

InChI

1S/C7H16N2O/c1-10-7-6-9-4-2-8-3-5-9/h8H,2-7H2,1H3

InChI key

BMEMBBFDTYHTLH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of biologically and pharmacologically active molecules

Warning

Air-sensitive - absorbs CO2

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1

Target Organs

respiratory tract irritation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service