Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70074

Sigma-Aldrich

Pullulan

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
Pricing and availability is not currently available.

form

powder

mol wt

Mw ~100000

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marieta Constantin et al.
Carbohydrate polymers, 91(1), 74-84 (2012-10-10)
Pullulan-graft-poly(3-acrylamidopropyl trimethylammonium chloride) (P-g-pAPTAC) microspheres were prepared by suspension cross-linking of the pullulan previously grafted with cationic moieties. Adsorption of Azocarmine B by the P-g-pAPTAC microspheres was used as a model to demonstrate the removal of anionic dyes from aqueous
Xiaojun Tao et al.
PloS one, 7(11), e49304-e49304 (2012-11-21)
Nanoparticle (NP) compositions such as hydrophobicity and surface charge are vital to determine the presence and amount of human serum albumin (HSA) binding. The HSA binding influences drug release, biocompatibility, biodistribution, and intercellular trafficking of nanoparticles (NPs). Here, we prepared
Yen Yen Chai et al.
Environmental technology, 33(10-12), 1231-1238 (2012-08-04)
Two thermophilic bacteria (SK3-4 and DT3-1) were isolated from the Sungai Klah (SK) and Dusun Tua (DT) hot springs in Malaysia. The cells from both strains were rod-shaped, stained Gram positive and formed endospores. The optimal growth of both strains
Anirban Roy Choudhury et al.
Microbial cell factories, 11(1), 39-39 (2012-04-03)
Ever increasing demand for fossil fuels is a major factor for rapid depletion of these non-renewable energy resources, which has enhanced the interest of finding out alternative sources of energy. In recent years jatropha seed oil has been used extensively
Shengjun Wu et al.
International journal of biological macromolecules, 55, 254-257 (2013-02-05)
Pullulan is a thickener that can form semipermeable films, and glutathione is an effective reducing agent, while chitooligosaccharide has antibacterial activity. In this study, effect of pullulan-based coatings in combination with antibrowning and antibacterial agents (1% pullulan; 0.8% glutathione+1% chitooligosaccharides;

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service