Skip to Content
MilliporeSigma
All Photos(1)

Documents

70028

Supelco

AQUANAL-professional SPECTRO 1000

10 cylindric tubes diameter 16 mm for Spectro 1000 (70012)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

application(s)

environmental
general analytical

Looking for similar products? Visit Product Comparison Guide

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service