Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69945

Sigma-Aldrich

3-Morpholino-1,2-propanediol

technical, ~90% (GC)

Synonym(s):

3-(4-Morpholinyl)-1,2-propanediol, 3-Morpholin-4-ylpropane-1,2-diol, N -(2,3-Dihydroxypropyl)morpholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO3
CAS Number:
Molecular Weight:
161.20
Beilstein:
107210
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical

Assay

~90% (GC)

refractive index

n20/D 1.493

bp

191 °C/30 mmHg (lit.)

mp

37-38 °C (lit.)

density

1.157 g/mL at 25 °C (lit.)

SMILES string

OCC(O)CN1CCOCC1

InChI

1S/C7H15NO3/c9-6-7(10)5-8-1-3-11-4-2-8/h7,9-10H,1-6H2

InChI key

VZBNUEHCOOXOHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service