Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69643

Sigma-Aldrich

2-Methylvaleric acid

purum, ≥98.0% (GC)

Synonym(s):

(±)-2-Methylvaleric acid, 2-Methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(CH3)CO2H
CAS Number:
Molecular Weight:
116.16
Beilstein:
1720658
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.414 (lit.)
n20/D 1.414

bp

196-197 °C (lit.)

density

0.931 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)C(O)=O

InChI

1S/C6H12O2/c1-3-4-5(2)6(7)8/h5H,3-4H2,1-2H3,(H,7,8)

InChI key

OVBFMEVBMNZIBR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoqing Cai et al.
Bioorganic & medicinal chemistry, 19(12), 3831-3844 (2011-05-27)
Previous studies have indicated that the methylvalerate subunit of bleomycin (BLM) plays an important role in facilitating DNA cleavage by BLM and deglycoBLM. Eleven methylvalerate analogues have been synthesized and incorporated into deglycoBLM congeners by the use of solid-phase synthesis.
Z Suarez de Mata et al.
Archives of biochemistry and biophysics, 285(1), 158-165 (1991-02-15)
The condensation of two propionyl-CoA units or a propionyl-CoA with acetyl-CoA is required for the synthesis of 2-methylvalerate or 2-methylbutyrate, respectively, two of the major fermentation products of Ascaris anaerobic muscle metabolism. An enzyme that preferentially catalyzes the condensation of
R Komuniecki et al.
The Journal of parasitology, 67(6), 841-846 (1981-12-01)
Disrupted Ascaris mitochondria formed 2-methylbutyrate (2-MB) and 2-methylvalerate (2-MV) when incubated anaerobically with acetyl CoA, propionyl CoA and NADH. However, when mitochondrial membranes were removed by high speed centrifugation and the mitochondrial soluble fraction was incubated with the same substrates
P R Komuniecki et al.
Molecular and biochemical parasitology, 22(2-3), 241-248 (1987-01-15)
Ascaris suum L3 larvae isolated from rabbit lungs undergo the third ecdysis to L4 larvae after 3 days in culture under a gas phase of 85% N2/10% CO2/5% O2. The L3 larvae contain substantial malic enzyme activity and are capable
Yu Meng et al.
Journal of bacteriology, 192(22), 5982-5990 (2010-09-21)
Short- and medium-chain acyl coenzyme A (acyl-CoA) synthetases catalyze the formation of acyl-CoA from an acyl substrate, ATP, and CoA. These enzymes catalyze mechanistically similar two-step reactions that proceed through an enzyme-bound acyl-AMP intermediate. Here we describe the characterization of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service