Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

695084

Sigma-Aldrich

Acetic acid

glacial, ≥99.85%

Synonym(s):

Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

vapor density

2.07 (vs air)

Assay

≥99.85%

autoignition temp.

800 °F

expl. lim.

16 %, 92 °F
4 %, 59 °F

impurities

≤0.0050% Nonvolatile matter
≤0.050% Formic acid
≤0.150% water
≤5 ppm Acetaldehyde

color

APHA: ≤5

refractive index

n20/D 1.371 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

freezing point ≥16.35 °C

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤1 ppm

cation traces

Fe: ≤0.50 ppm
heavy metals: ≤0.5 ppm

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeynab Yaghoubi et al.
Materials science & engineering. C, Materials for biological applications, 103, 109779-109779 (2019-07-28)
A novel MWCNT-PNIPAAm nanocomposite membrane was developed with an excellent cleaning efficiency of thermo-responsive surface. The thermo-responsive N-isopropyle acryleamide (NIPAAm) monomer was polymerized on the surface of MWCNT via free radical polymerization. The prepared MWCNT-PNIPAAm nanocomposite was characterized by FTIR
Isabelle Brunet et al.
The Journal of clinical investigation, 124(7), 3230-3240 (2014-06-18)
Autonomic sympathetic nerves innervate peripheral resistance arteries, thereby regulating vascular tone and controlling blood supply to organs. Despite the fundamental importance of blood flow control, how sympathetic arterial innervation develops remains largely unknown. Here, we identified the axon guidance cue
Katharine Trenholme et al.
Antimicrobial agents and chemotherapy, 58(7), 3666-3678 (2014-04-16)
Therapies to prevent transmission of malaria parasites to the mosquito vector are a vital part of the global malaria elimination agenda. Primaquine is currently the only drug with such activity; however, its use is limited by side effects. The development
Grete Andersen et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 308(2), R123-R130 (2014-11-21)
In healthy individuals, postexercise protein supplementation increases muscle protein anabolism. In patients with muscular dystrophies, aerobic exercise improves muscle function, but the effect of exercise on muscle protein balance is unknown. Therefore, we investigated 1) muscle protein balance before, during
Ilya Chumakov et al.
Orphanet journal of rare diseases, 9, 201-201 (2014-12-11)
Charcot-Marie-Tooth disease type 1A (CMT1A) is the most common inherited sensory and motor peripheral neuropathy. It is caused by PMP22 overexpression which leads to defects of peripheral myelination, loss of long axons, and progressive impairment then disability. There is no

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service