Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69246

Supelco

Mineral oil standard mixture Type A and B for EN 14039 and ISO 16703

analytical standard, ~8 mg/mL in heptane (exact content on the label)

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$68.20
10 ML
$202.00

$68.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 ML
$68.20
10 ML
$202.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$68.20


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

100
200

Agency

EN 14039
ISO 16703

shelf life

limited shelf life, expiry date on the label

concentration

~8 mg/mL in heptane (exact content on the label)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.686 g/mL at 20 °C

application(s)

environmental
petroleum

format

multi-component solution

Analysis Note

Contains Mineral oil type A and B diluted with n-heptane; contains n-decane and n-tetracontane as internal standards.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Proceedings of World Academy of Science, Engineering and Technology, 3 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service