Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

69193

Supelco

Methyl nicotinate

analytical standard

Synonym(s):

Nicotinic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
Beilstein:
113951
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

white to off-white

bp

204 °C (lit.)

mp

39-42 °C
42-44 °C (lit.)

solubility

H2O: 10%, clear to very faintly turbid, colorless

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COC(=O)c1cccnc1

InChI

1S/C7H7NO2/c1-10-7(9)6-3-2-4-8-5-6/h2-5H,1H3

InChI key

YNBADRVTZLEFNH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl nicotinate is a volatile product of certain fruits such as carambola, soursop, wild strawberries, etc. It is used for a variety of applications in food products, cosmetics, and medicinal products.

Application

Methyl nicotinate may be used as a test compound to study the percutaneous penetration of methyl nicotinate through human skin in vitro and in vivo using micro-dialysis technique and different analytical techniques.
Methyl nicotinate may be used as an analytical reference standard for the determination of the analyte in biological samples, cosmetics, and pharmaceuticals by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC-quantification of diethylamine salicylate and methyl nicotinate in ointments
Abounassif MA, et al.
Journal of Liquid Chromatography and Related Technologies, 15(4), 625-636 (1992)
Determination of methyl nicotinate in pharmaceutical creams by high-performance thin-layer chromatography
De Spiegeleer B, et al.
Chromatographia, 20(4), 249-252 (1985)
Determination of nicotinic acid and metabolites in urine, cosmetics, and pharmaceuticals by high-performance liquid chromatography with novel electrochemical detection
Wang L-H
Analytical Letters, 49(10), 1467-1479 (2016)
Microdialysis technique as a method to study the percutaneous penetration of methyl nicotinate through excised human skin, reconstructed epidermis, and human skin in vivo
Boelsma E, et al.
Pharmaceutical Research, 17(2), 141-147 (2000)
Active Ingredients Used in Cosmetics: Safety Survey
Health protection of consumers (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service