Skip to Content
MilliporeSigma
All Photos(2)

Documents

68814

Supelco

D-α-Tocotrienol

analytical standard

Synonym(s):

(R)-α-Tocotrienol, [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

faint beige to beige
white to yellow

application(s)

vitamins, nutraceuticals, and natural products

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\CC[C@]1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H44O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h12,14,16,30H,9-11,13,15,17-19H2,1-8H3/b21-14+,22-16+/t29-/m1/s1

InChI key

RZFHLOLGZPDCHJ-XZXLULOTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Tocotrienol is one of the lipid-soluble isomers of the essential micronutrient vitamin E, which exhibits potent antioxidant performance in lipoproteins and polyunsaturated fatty acids by exerting efficient free radical scavenging action.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive analysis of vitamin E constituents in human plasma by liquid chromatography- mass spectrometry
Nagy K, et al.
Analytical Chemistry, 79(18), 7087-7096 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service