Skip to Content
MilliporeSigma
All Photos(1)

Documents

68630

Sigma-Aldrich

(N-Methylphenylamino)triphenylphosphonium iodide

purum, ≥98.0% (NT)

Synonym(s):

Murahashi reagent, N-Methylanilinotriphenylphosphonium iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23INP
CAS Number:
Molecular Weight:
495.34
Beilstein:
4075159
MDL number:
UNSPSC Code:
12352103

grade

purum

Assay

≥98.0% (NT)

mp

237-242 °C (dec.)

SMILES string

[I-].CN(c1ccccc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

Other Notes

Reagent used for the activation of alcohols; reaction of the active intermediate with amines, thiols and azides to the respective derivatives; C-Alkylation or C-arylation of allylic alcohols

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Tanigawa et al.
Journal of the American Chemical Society, 99, 2361-2361 (1977)
H.L. Goering et al.
The Journal of Organic Chemistry, 50, 1597-1597 (1985)
Tetrahedron Letters, 27, 6353-6353 (1986)
H.L. Goering et al.
The Journal of Organic Chemistry, 46, 2144-2144 (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service