Skip to Content
MilliporeSigma
All Photos(1)

Documents

68589

Millipore

Brilliant Green + Novobiocin Selective Supplement

suitable for microbiology

Synonym(s):

Muller-Kauffmann Tetrathionate Broth Supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

suitability

Salmonella spp.

Application

Novobiocin+Brillant green selective supplement is added to Muller-Kauffmann Tetrationate medium base in order to obtain a complete medium for the enrichment of enteric or intestinal pathogens, and for all the members of Salmonella type.
Usually this medium is used for the analysis of polluted samples, like faeces, urine, waste water and others.
MKTTn was developed by Muller and later modified by Kauffmann with the addition of ox bile and brilliant green to improve selectivity. The addition of novobiocin was later described by Jeffries to improve inhibition of Proteus species.

Components

Compositon (per vial, sufficient for 500 ml medium)
Novobiocin 20mg
Brilliant Green 5 mg

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service