Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

68488

Supelco

Phenethyl isothiocyanate

analytical standard

Synonym(s):

PEITC, 2-Phenylethyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2NCS
CAS Number:
Molecular Weight:
163.24
Beilstein:
2084162
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.5888 (lit.)
n20/D 1.589-1.591

bp

139-140 °C/11 mmHg (lit.)

density

1.094 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

S=C=NCCc1ccccc1

InChI

1S/C9H9NS/c11-8-10-7-6-9-4-2-1-3-5-9/h1-5H,6-7H2

InChI key

IZJDOKYDEWTZSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phenethyl isothiocyanate, an organic isothiocyanate, is a cruciferous vegetable ingredient present as a glucosinolate in cabbage, turnips, radishes, etc. It is a well-known chemopreventive agent reported to be effective against cancers affecting the lung, breast, pancreas, prostate and so on.

Application

Phenethyl isothiocyanate may be used as an analytical standard for the determination of the analyte in biological samples and vegetable extracts by various chromatography techniques.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Negrusz et al.
Journal of chromatography. B, Biomedical sciences and applications, 718(1), 193-198 (1998-12-01)
Phenethyl isothiocyanate is unstable in aqueous media and at low pH, and rapidly degrades to phenethylamine. Concentrations of phenethylamine, a phenethyl isothiocyanate marker, in dog plasma, were determined utilizing solid-phase extraction and gas chromatography-mass spectrometry with chemical ionization using acetone
L Liebes et al.
Analytical biochemistry, 291(2), 279-289 (2001-06-13)
Dietary and pharmacologic isothiocyanates (ITCs) may play a role in reducing the risk of certain cancers. The quantification of ITCs in humans is important both for epidemiological and pharmacokinetic studies. We describe a modification of an HPLC-based assay of urinary
Stefan Tyroller et al.
Toxicology, 215(3), 245-253 (2005-08-25)
N'-Nitrosonornicotine (NNN) was the first tobacco-specific nitrosamine (TSNA) identified as carcinogen in tobacco smoke, but no data exist on in vivo interactions between NNN and other tobacco alkaloids, TSNA or phenethyl isothiocyanate (PEITC) which have been demonstrated in various studies
Yan Ji et al.
Analytical biochemistry, 323(1), 39-47 (2003-11-19)
Phenethyl isothiocyanate (PEITC) is a dietary compound present in cruciferous vegetables that has cancer-preventive properties. Our objective was to develop and validate a novel liquid chromatography-tandem mass spectrometry procedure to analyze PEITC concentrations in human plasma and urine. Following hexane
Lijiang Song et al.
Analytical biochemistry, 347(2), 234-243 (2005-11-18)
Dietary glucosinolates are under intensive investigation as precursors of cancer-preventive isothiocyanates. Quantitation of the dose and bioavailability of glucosinolates and isothiocyanates requires a comprehensive analysis of the major dietary glucosinolates, isothiocyanates, and related metabolites. We report a liquid chromatography with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service