Skip to Content
MilliporeSigma
All Photos(1)

Documents

68353

Supelco

DL-Serine

analytical standard

Synonym(s):

(±)-2-Amino-3-hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
105.09
Beilstein:
1721405
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (TLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

color

white to off-white

mp

240 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

NC(CO)C(O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Pablo M Palavecino et al.
Journal of the science of food and agriculture, 99(3), 1351-1357 (2018-08-11)
The development of new products with a focus on nutrition, rather than other technical aspects, is essential to improve the quality of celiac diets. Nutritional attributes of white and brown sorghum gluten-free pasta developed in a previous work were analyzed.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service