Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

67579

Millipore

O157 Millichrome plus Agar

suitable for microbiology

Synonym(s):

E. coli O157 chromogenic agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41161501
NACRES:
NA.74

Agency

EP
JP
USP

Quality Level

Sterilization Compatibility

dry heat compatible

form

dry powder

shelf life

limited shelf life, expiry date on the label

composition

Agar, 15.0 g/L
Chromogenic Mix, 1.2 g/L
Chromogenic mix, 1.2 g/L
Peptone and yeast
extract, 13.0 g/L

Peptone & Yeast Extract, 13.0 g/L

manufacturer/tradename

MilliChrome

technique(s)

microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

solubility

deionized water: 29.2 g/L

application(s)

food and beverages
microbiology

shipped in

ambient

storage temp.

15-25°C

suitability

selective for bacteria: Escherichia coli O157

General description

O157 Millichrome plus Agar is a selective media for the isolation and detection of E. coli O157. The distinguishing feature of these media is the inclusion of a chromogenic substrate mix that produces visible color changes upon enzymatic activity of E. coli O157. Millichrome chromogenic media is a specialized type of culture medium used to detect and isolate specific groups of bacteria based on their ability to produce enzymes with chromogenic substrates. For optimal growth, peptone and yeast extract provide nitrogenous nutrients and essential growth factors. Sodium chloride is crucial for maintaining osmotic balance. The chromogenic mix contains substrates that change color when characteristic enzymes cleave them, allowing for easy differentiation.To create a more selective and specific medium, it is recommended to include a potassium tellurite supplement. If there is expected to be a high load of Proteus spp., cefixime can be added. Similarly, a high load of Pseudomonas and/or Aeromonas can be inhibited by including cefsulodin. Agar serves as the solidifying agent.

Application

O157 Millichrome plus Agar Base is a selective chromogenic culture medium intended for use in the qualitative direct detection, differentiation and presumptive identification of pathogenic biotypes of E. coli O157 from various samples.

Preparation Note

Step 1 (Preparation)
  • Disperse slowly 29.2 g of powder base in 1 L of purified water.
  • Stir until agar is well thickened.
  • Heat and bring to boil (100 °C) while swirling or stirring regularly.
DO NOT HEAT TO MORE THAN 100 °C. DO NOT AUTOCLAVE AT 121 °C.
Warning: If using an autoclave, do so without pressure.

Advice 1: For the 100 °C heating step, mixture may also be brought to a boil in a microwave oven: after initial boiling, remove from oven, stir gently, then return to oven for short repeated bursts of heating until complete fusion of the agar grains has taken place (large bubbles replacing foam).
Advice 2: if a more selective, and more specific, medium is needed, add a solution of Potassium Tellurite to obtain a final concentration of 2.5 mg/L at 45-50 °C.
Advice 3: in case of product samples containing a high load of Proteus, Cefixime can be added at 0.025 mg/L at 45-50 °C.
Advice 4 : in case of product samples containing a high load of Pseudomonas and/or Aeromonas, Cefsulodin can be added at 5 mg/L at 45-50 °C.

Step 2 (Pouring plates)
  • Cool in a water bath at 45-50 °C, swirling or stirring gently.
  • Pour into sterile Petri dishes.
  • Let it solidify and dry.

Storage and Stability

Store prepared media below 8°C, protected from direct light (max. 1 month).Store dehydrated powder, in a dry place, in tightly sealed containers at 2-25°C.

Analysis Note

The E. coli O157 is detected by a characteristic mauve color after only 24 h of incubation at an optimal growth temperature of 37°C, while most other E. coli form blue color colonies after incubation at the same temperature

Legal Information

MilliChrome is a trademark of Merck KGaA, Darmstadt, Germany

supplement

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selective media enable faster results and visual confirmation for the detection, identification, and enumeration of microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service