Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67135

Supelco

Isovitexin

analytical standard

Synonym(s):

6-C-Glucosylapigenin, Homovitexin, Saponaretin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
Beilstein:
66651
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤12.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)cc4

InChI

1S/C21H20O10/c22-7-14-17(26)19(28)20(29)21(31-14)16-11(25)6-13-15(18(16)27)10(24)5-12(30-13)8-1-3-9(23)4-2-8/h1-6,14,17,19-23,25-29H,7H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

MYXNWGACZJSMBT-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isovitexin is one of the main flavone C-glycosides and a major constituent found in pigeon pea leaves. It exhibits antibacterial and anti-inflammatory activities, inhibition of capillary permeability and abirritative properties.[1]

Application

Isovitexin may be used as an analytical reference standard for the separation and determination of the analyte in medicinal plant extracts[1][2] using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Phytochemical found in medicinal herbs. Antioxidant.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

An improved HPLC determination of flavonoids in medicinal plant extracts
Pietta.G.P, et al.
Chromatographia, 27(9-10), 509-512 (1989)
Jong Min Kim et al.
Chemistry & biodiversity, 5(2), 352-356 (2008-02-23)
Ellagic acid (1), 3,3'-di-O-methylellagic acid (2), 3,3',4-tri-O-methylellagic acid (3), isovitexin (4), kaempferol 3-O-beta-D-glucuronide methyl ester (5), quercetin 3-O-alpha-L-arabinopyranosyl-(1-->6)-beta-D-galactopyranoside (6), ursolic acid, pomolic acid, tormentic acid, euscaphic acid, euscaphic acid 28-O-beta-D-glucopyranoside, and maslinic acid were isolated from the AcOEt- and BuOH-soluble
C Y Choo et al.
Journal of ethnopharmacology, 142(3), 776-781 (2012-06-12)
The leaves of Ficus deltoidea are used as a traditional medicine by diabetes patients in Malaysia. The objective of the study is to identify and evaluate bioactive constituents with in vivo α-glucosidase inhibition. The partitioned extracts, subfractions and pure bioactive
Yujie Fu et al.
Journal of separation science, 31(2), 268-275 (2008-01-16)
A method based on ultrasonic extraction (USE) followed by LC-MS is presented for the determination of vitexin and isovitexin in pigeonpea extracts in this study. The influential parameters of the USE procedure were optimized, and the optimal conditions were as
Yujie Fu et al.
Journal of chromatography. A, 1139(2), 206-213 (2006-12-05)
Vitexin and isovitexin are a pair of isomeric compounds known as the major constituents in pigeonpea leaves and possess various pharmacological activities. In the present study, the preparative separation of vitexin and isovitexin with macroporous resins (Nankai Hecheng S &

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service