Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

65958

Sigma-Aldrich

Bromomethane-13C

purum, 99 atom % 13C, cylinder (incl. valve, net weight ~2 g)

Synonym(s):

Methyl-13C bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH3Br
CAS Number:
Molecular Weight:
95.93
Beilstein:
2231126
MDL number:
UNSPSC Code:
12352200

vapor density

3.3 (20 °C, vs air)

Quality Level

vapor pressure

1420 mmHg ( 20 °C)

isotopic purity

99 atom % 13C

grade

purum

form

cylinder (incl. valve, net weight ~2 g)

autoignition temp.

996 °F

expl. lim.

16 %

bp

4 °C (lit.)

mp

−94 °C (lit.)

density

1.73 g/mL at 0 °C (lit.)

mass shift

M+1

SMILES string

[13CH3]Br

InChI

1S/CH3Br/c1-2/h1H3/i1+1

InChI key

GZUXJHMPEANEGY-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Sure/Pac® cylinder, Recommended valves see section ′Tools/Valves and Adapter for Cylinders′ in the annex at the end of this catalog.

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Muta. 2 - Ozone 1 - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service