Skip to Content
MilliporeSigma
All Photos(1)

Documents

65200

Sigma-Aldrich

4-Methoxyphenylacetic acid

purum, ≥98.0% (T)

Synonym(s):

Homoanisic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
166.17
Beilstein:
1101737
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

grade

purum

Assay

≥98.0% (T)

bp

140 °C/3 mmHg (lit.)

mp

84-86 °C (lit.)
84-86 °C

SMILES string

COc1ccc(CC(O)=O)cc1

InChI

1S/C9H10O3/c1-12-8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)

InChI key

NRPFNQUDKRYCNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service