Skip to Content
MilliporeSigma
All Photos(1)

Documents

64008BC

Millipore

AssureSwab Environmental Sampling

BioControl, Self-contained environmental sampling device for use in environmental monitoring

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112601

shelf life

6‑18 mo.

manufacturer/tradename

BioControl Systems 64008-100

application(s)

food and beverages

storage temp.

2-8°C

General description

The AssureSwab is a self-contained environmental sampling device for use in environmental monitoring. The AssureSwab sampling device consists of a Dacron tipped swab and letheen broth to facilitate recovery of bacteria. It can be used with most film and plating systems including the SimPlate method. The AssureSwab sampling device can be used on either wet or dry surfaces to monitor the processing environment as part of a HACCP monitoring program.

Components

100 Sampling devices

Kit Components Also Available Separately

Product No.
Description
SDS

  • Dry Chamber

  • Wet Chamber

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service