Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63130

Sigma-Aldrich

Magnesium peroxide light

purum p.a., MgO2.(MgO)n, 24-28% MgO2 basis (RT)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

reaction suitability

reagent type: oxidant

concentration

24-28% (MgO2, RT)

anion traces

chloride (Cl-): ≤200 mg/kg
sulfate (SO42-): ≤2000 mg/kg

cation traces

Ca: ≤5000 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
K: ≤100 mg/kg
Na: ≤500 mg/kg
Ni: ≤100 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Mg++].[O-][O-]

InChI

1S/Mg.O2/c;1-2/q+2;-2

InChI key

SPAGIJMPHSUYSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service