Skip to Content
MilliporeSigma
All Photos(2)

Documents

63088

Supelco

Magnesium ionophore IV

Selectophore, function tested

Synonym(s):

ETH 7025, N,N′,N′′-Tris[3-(heptylmethylamino)-3-oxopropionyl]-8,8′-iminodioctylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H94N6O6
CAS Number:
Molecular Weight:
863.31
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

liquid

quality

function tested

SMILES string

CCCCCCCN(C)C(=O)CC(=O)NCCCCCCCCN(CCCCCCCCNC(=O)CC(=O)N(C)CCCCCCC)C(=O)CC(=O)N(C)CCCCCCC

InChI

1S/C49H94N6O6/c1-7-10-13-22-29-36-52(4)46(58)41-44(56)50-34-27-20-16-18-25-32-39-55(49(61)43-48(60)54(6)38-31-24-15-12-9-3)40-33-26-19-17-21-28-35-51-45(57)42-47(59)53(5)37-30-23-14-11-8-2/h7-43H2,1-6H3,(H,50,56)(H,51,57)

InChI key

JYBGTBZZVHYIJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples
Very lipophilic magnesium ionophore; assay of Mg2+-activity in human blood serum with an optimized liquid polymer membrane electrode

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Physical form

50 mg solution in 0.5 mL tetrahydrofuran

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Free Intracellular Magnesium Remains Uninfluenced by Changes of Extracellular Magnesium in Cardiac Guinea Pig Papillary Muscle
S. Gasser et al.
Journal of Clinical and Basic Cardiology, 8, 29-32 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service