61848
Isovaleraldehyde
analytical standard
Synonym(s):
3-Methylbutanal, 3-Methylbutyraldehyde, NSC 404119
Select a Size
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
2.96 (vs air)
vapor pressure
30 mmHg ( 20 °C)
Assay
≥93.5% (GC)
autoignition temp.
464 °F
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤1% isovaleric acid
refractive index
n20/D 1.388 (lit.)
bp
90 °C (lit.)
density
0.803 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
Organoleptic
pungent
storage temp.
2-8°C
SMILES string
[H]C(=O)CC(C)C
InChI
1S/C5H10O/c1-5(2)3-4-6/h4-5H,3H2,1-2H3
InChI key
YGHRJJRRZDOVPD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
32.9 °F - closed cup
Flash Point(C)
0.5 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
-Tolualdehyde; Valeraldehyde; Isovaleraldehyde
Chromatograms
suitable for GC, application for SPMEActive Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service