Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61237

Supelco

Frangulin A

analytical standard

Synonym(s):

3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-1,8-dihydroxy-6-methylanthraquinone, Emodin-L-rhamnoside, Franguloside, Rhamnoxanthin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O9
CAS Number:
Molecular Weight:
416.38
Beilstein:
59993
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water (Karl Fischer)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](OC2=CC(O)=C3C(C(C(C=C(C)C=C4O)=C4C3=O)=O)=C2)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C21H20O9/c1-7-3-10-14(12(22)4-7)18(26)15-11(17(10)25)5-9(6-13(15)23)30-21-20(28)19(27)16(24)8(2)29-21/h3-6,8,16,19-24,27-28H,1-2H3/t8-,16-,19+,20+,21-/m0/s1

InChI key

DTTVUKLWJFJOHO-FUCRAMRQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiying Liu et al.
Journal of neurochemistry, 152(3), 397-415 (2019-08-24)
Parkinson's disease (PD) is the second most common neurodegenerative disease after Alzheimer's disease. Its pathological features are dopaminergic neuronal death in the substantia nigra (SN), and significant reduction in dopamine (DA) content in the striatum. A large number of studies

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service