Skip to Content
MilliporeSigma
All Photos(1)

Documents

611735

Supelco

Triphenyl phosphate solution

NMR reference standard, 0.0485 M in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.24

grade

NMR reference standard
analytical standard

Quality Level

concentration

0.0485 M in chloroform-d (99.8 atom % D)

technique(s)

NMR: suitable

NMR tube size

10 mm × 8 in.

application(s)

environmental

format

single component solution

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

31P sensitivity

Quantity

10 mm O.D. tube contains 3.175 mL

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory
Johanna Jyrkkärinne et al.
Journal of medicinal chemistry, 51(22), 7181-7192 (2008-11-06)
The human constitutive androstane receptor (CAR, NR1I3) is an important regulator of xenobiotic metabolism and other physiological processes. So far, only few CAR agonists are known and no explicit mechanism has been proposed for their action. Thus, we aimed to
Katsumasa Abe et al.
Applied and environmental microbiology, 80(18), 5866-5873 (2014-07-20)
Phosphotriesterases catalyze the first step of organophosphorus triester degradation. The bacterial phosphotriesterases purified and characterized to date hydrolyze mainly aryl dialkyl phosphates, such as parathion, paraoxon, and chlorpyrifos. In this study, we purified and cloned two novel phosphotriesterases from Sphingomonas
Blagoj Mitrevski et al.
Journal of chromatography. A, 1362, 262-269 (2014-09-03)
Quadrupole time-of-flight mass spectrometry (Q-TOFMS) has been evaluated with respect to its applicability in comprehensive two-dimensional gas chromatography (GC×GC). At a maximum acquisition frequency, while approximately 50 full accurate mass spectra on disk were acquired per s (50Hz) in scan
Zulin Zhang et al.
Journal of separation science, 37(24), 3699-3705 (2014-10-02)
A simple and rapid method was developed for the simultaneous analysis of nine different pesticides in water samples by gas chromatography with mass spectrometry. A number of parameters that may affect the recovery of pesticides, such as the type of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service