Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60441

Sigma-Aldrich

Potassium perchlorate

puriss. p.a., ACS reagent, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KClO4
CAS Number:
Molecular Weight:
138.55
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Assay

≥99.0%

impurities

≤0.0005% total nitrogen (N)

pH

5.0-7.0 (25 °C, 1% in H2O)

mp

400 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤30 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].[O-]Cl(=O)(=O)=O

InChI

1S/ClHO4.K/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

YLMGFJXSLBMXHK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Ox. Sol. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Hiasa et al.
Japanese journal of cancer research : Gann, 78(12), 1335-1340 (1987-12-01)
The effect of 1000 ppm potassium perchlorate (KClO4), 1000 ppm potassium iodide (KI) or 1000 ppm propylthiouracil (PTU) in the diet on the development of thyroid tumors was studied histologically and biochemically in Wistar rats given a single ip injection
J D Méndez-Díaz et al.
Water research, 43(6), 1621-1629 (2009-01-17)
The present study investigates the efficacy of various oxidizing treatments (ClO(-), ClO(2), KMnO(4), O(3), O(3)/H(2)O(2), O(3)/activated carbon) to remove from waters sodium dodecylbenzenesulphonate (SDBS), considered as model surfactant. Results obtained show that the use of ClO(-) and ClO(2) does not
G Coelho-Palermo Cunha et al.
Regulatory toxicology and pharmacology : RTP, 48(3), 270-278 (2007-06-19)
The perchlorate discharge assay (PDA) is potentially of high diagnostic value to distinguish between direct and indirect thyroid toxicity mechanisms, provided that standard treatment times are established and positive controls yield reproducible results. Therefore the PDA was evaluated after 2
F Bogazzi et al.
Journal of endocrinological investigation, 29(1), 61-66 (2006-03-24)
Amiodarone perturbs thyroid function, causing overt hypothyroidism or hyperthyroidism in 15% of patients. Changes in thyroid function are likely due, at least in part, to amiodarone and/or desethylamiodarone (DEA) concentration into the thyroid gland, but mechanisms whereby the drug uptake
Benedicte Thienpont et al.
Environmental science & technology, 45(17), 7525-7532 (2011-08-02)
Thyroxine-immunofluorescence quantitative disruption test (TIQDT) was designed to provide a simple, rapid, alternative bioassay for assessing the potential of chemical pollutants and drugs to disrupt thyroid gland function. This study demonstrated that zebrafish eleutheroembryos provided a suitable vertebrate model, not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service