Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60338

Sigma-Aldrich

Potassium bicarbonate

puriss. p.a., ACS reagent, ≥99.7% (calc. based on dry substance, T)

Synonym(s):

Potassium hydrogen carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Beilstein:
4535309
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Assay

≥99.7% (calc. based on dry substance, T)

impurities

≤0.003% sulfur compounds (as SO4)

anion traces

chloride (Cl-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤5 mg/kg
Na: ≤200 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[K+].OC([O-])=O

InChI

1S/CH2O3.K/c2-1(3)4;/h(H2,2,3,4);/q;+1/p-1

InChI key

TYJJADVDDVDEDZ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lisa Ceglia et al.
The Journal of clinical endocrinology and metabolism, 94(2), 645-653 (2008-12-04)
Protein is an essential component of muscle and bone. However, the acidic byproducts of protein metabolism may have a negative impact on the musculoskeletal system, particularly in older individuals with declining renal function. We sought to determine whether adding an
Fj Wegehaupt et al.
Oral diseases, 17(5), 508-514 (2011-02-22)
The aim of this study was to evaluate the erosive/abrasive enamel wear after contact with orange juices modified with different dietary supplements. A total of 96 bovine enamel samples were prepared and allocated to eight groups (1-8; n = 12).
K Holtenius et al.
Journal of dairy science, 91(2), 743-748 (2008-01-26)
The objective of the present study was to investigate the interactions of dietary K intake typical for forage-based diets on Mg balance in lactating dairy cows. Six lactating multiparous cows of the Swedish Red and White breed in midlactation were
Karalee Jarvis et al.
Environmental science & technology, 43(16), 6314-6319 (2009-09-15)
Storage of CO2 through mineral sequestration using olivine has been shown to produce environmentally benign carbonates. However, due to the formation of a rate-limiting reaction product layer, the rate of reaction is insufficient for large-scale applications. We report the results
Daniel L Belavý et al.
Journal of applied physiology (Bethesda, Md. : 1985), 111(5), 1304-1314 (2011-07-30)
As part of the nutrition-countermeasures (NUC) study in Cologne, Germany in 2010, seven healthy male subjects underwent 21 days of head-down tilt bed rest and returned 153 days later to undergo a second bout of 21-day bed rest. As part

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service