Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60313

Sigma-Aldrich

Potassium hexacyanocobalt(II)-ferrate(II)

purum p.a., ≥97.0% (KT)

Synonym(s):

Dipotassium hexacyanocobalt(II)-ferrate(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2[CoFe(CN)6]
CAS Number:
Molecular Weight:
349.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

purum p.a.

Assay

≥97.0% (KT)

particle size

30-60 mesh

SMILES string

[K+].[K+].N#C[Fe-](C#N)C#N.N#C[Co-](C#N)C#N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Compound used for the ion-exchange absorption of cesium ions

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.E. Prout et al.
J. Inorg. Nucl. Chem., 27, 473-473 (1965)
P. Nielsen et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 42, 1451-1451 (1987)
J. Nagy et al.
Isotopenpraxis, 20, 101-101 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service