Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58351

Sigma-Aldrich

2-Methylpropane

puriss.

Synonym(s):

Isobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CH
CAS Number:
Molecular Weight:
58.12
Beilstein:
1730720
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

2.01 (21 °C, vs air)

vapor pressure

72.2 psi ( 37.7 °C)

grade

puriss.

autoignition temp.

860 °F

expl. lim.

8.3 %

bp

−12 °C (lit.)

mp

−160 °C (lit.)

density

2.064 g/mL at 25 °C (lit.)

SMILES string

CC(C)C

InChI

1S/C4H10/c1-4(2)3/h4H,1-3H3

InChI key

NNPPMTNAJDCUHE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin with 250 mL (net ~140 g)

Other Notes

Sales restrictions may apply

control valve

Product No.
Description
Pricing

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1A - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-117.4 °F - closed cup

Flash Point(C)

-83 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vera Lepperhof et al.
PloS one, 9(9), e107363-e107363 (2014-09-17)
Cell loss after transplantation is a major limitation for cell replacement approaches in regenerative medicine. To assess the survival kinetics of induced pluripotent stem cell (iPSC)-derived cardiomyocytes (CM) we generated transgenic murine iPSC lines which, in addition to CM-specific expression
Christian Geis et al.
Experimental neurology, 265, 8-21 (2014-12-30)
Neuromyelitis optica (NMO) is causally linked to autoantibodies (ABs) against aquaporin 4 (AQP4). Here, we focused on the pathogenic effects exclusively mediated by human ABs to AQP4 in vivo. We performed cell-free intrathecal (i.th.) passive transfer experiments in Lewis rats

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service