Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

56677

Supelco

Formaldehyde-2,4-DNPH

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Formaldehyde-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$209.00

$209.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$209.00

About This Item

Linear Formula:
H2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
210.15
Beilstein:
750330
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$209.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage condition

under inert gas (Argon)

technique(s)

gas chromatography (GC): suitable

mp

153-156 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1ccc(NN=C)c(c1)[N+]([O-])=O

InChI

1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2

InChI key

UEQLSLWCHGLSML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Amine-impregnated dendritic mesoporous silica for formaldehyde adsorption: A study demonstrated the use of amine-impregnated dendritic mesoporous silica for effective formaldehyde adsorption, showcasing its potential in enhancing environmental safety and occupational health in settings with volatile formaldehyde emissions (Lee et al., 2023).

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicola Mucci et al.
La Medicina del lavoro, 110(6), 446-458 (2019-12-18)
In recent years, under-vacuum sealing (UVS) and containers with formalin encapsulated in the lid have been proposed for the reduction of occupational exposure to airborne formaldehyde (FA) in healthcare environments. We are presenting a study focused on the assessment of
Ikuo Ueta et al.
Analytical and bioanalytical chemistry, 407(3), 899-905 (2014-09-10)
A novel miniaturized sample extraction capillary was developed to provide a simple and sensitive method for analyzing gaseous formaldehyde (FA) using conventional high-performance liquid chromatography (HPLC). The extraction capillary was prepared by packing silica gel particles in a stainless steel

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. 저장조건이 under inert gas (Argon)로 되어있는데, 어떤 방식으로 보관하는 걸까요? 저장 조건이 room temperature인 다른 표준품처럼 보관하면 문제가 되는걸까요?

    1 answer
    1. This product should be stored between 2-8°C under an inert gas such as argon.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service