Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

56614

Supelco

Michler′s ketone

certified reference material, TraceCERT®

Synonym(s):

4,4′-Bis(dimethylamino)benzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NC6H4]2CO
CAS Number:
Molecular Weight:
268.35
Beilstein:
790733
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

174-176 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

CN(C)c1ccc(cc1)C(=O)c2ccc(cc2)N(C)C

InChI

1S/C17H20N2O/c1-18(2)15-9-5-13(6-10-15)17(20)14-7-11-16(12-8-14)19(3)4/h5-12H,1-4H3

InChI key

VVBLNCFGVYUYGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Dam. 1 - Muta. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service