Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

56291

Supelco

Clarithromycin N-oxide

analytical standard

Synonym(s):

6-O-Methylerythromycin N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H69NO14
CAS Number:
Molecular Weight:
763.95
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

O[C@@H]([C@@](O)(C)[C@@H](CC)OC([C@H](C)[C@H]1O[C@@H]2O[C@@H](C)[C@H](O)[C@](C)(OC)C2)=O)[C@@H](C)C([C@H](C)C[C@@](C)(OC)[C@H](O[C@@H]3O[C@H](C)C[C@H]([N+](C)([O-])C)[C@H]3O)[C@H]1C)=O

InChI

1S/C38H69NO14/c1-15-26-38(10,45)31(42)21(4)28(40)19(2)17-37(9,48-14)33(53-35-29(41)25(39(11,12)46)16-20(3)49-35)22(5)30(23(6)34(44)51-26)52-27-18-36(8,47-13)32(43)24(7)50-27/h19-27,29-33,35,41-43,45H,15-18H2,1-14H3/t19-,20-,21+,22+,23-,24+,25+,26-,27+,29-,30+,31-,32+,33-,35+,36-,37-,38-/m1/s1

InChI key

KDRPFIWYMNONLJ-KCBOHYOISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service