55805
Piperyline
analytical standard
Synonym(s):
1-Piperoylpyrrolidine, Piperiline, Pyrroperine, Trichostachine
About This Item
Recommended Products
grade
analytical standard
Quality Level
Assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
food and beverages
format
neat
SMILES string
O=C(\C=C\C=C\c1ccc2OCOc2c1)N3CCCC3
InChI
1S/C16H17NO3/c18-16(17-9-3-4-10-17)6-2-1-5-13-7-8-14-15(11-13)20-12-19-14/h1-2,5-8,11H,3-4,9-10,12H2/b5-1+,6-2+
InChI key
GQIJYUMTOUBHSH-IJIVKGSJSA-N
Application
Recommended products
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service